Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution for biopharmaceutical production
Corrosive
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution for biopharmaceutical production
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Sodium hydroxide 0.5 mol/l (0.5 N) in aqueous solution for biopharmaceutical production
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    0.495 --> 0.505 mol/l
  • Appearance:
    Passes test
  • CO3 (as Na2CO3):
    Max. 0.2 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 30 ppm
  • Fe (Iron):
    Max. 3 ppm
  • Residual metal catalysts:
    Unlikely to be present
  • Residual solvents (ICH Q3C):
    Excl.by manufact. process
  • Endotoxin:
    Max. 2.0 EU/ml
  • Filtered through 0.2 μm:
    Confirmed

Specification Test Results

Frequently Bought Together