Order Entry
Belgium
ContactUsLinkComponent
Buffer, di-Sodium hydrogen phosphate solution 0,066 M, pH 9,2, Supelco®
Buffer, di-Sodium hydrogen phosphate solution 0,066 M, pH 9,2, Supelco®
Catalog # 1.06587.1000
Supplier:  Merck
CAS Number:  
undefined
Buffer, di-Sodium hydrogen phosphate solution 0,066 M, pH 9,2, Supelco®
Catalog # 1.06587.1000
Supplier:  Merck
CAS Number:  

Specifications

  • pH buffer type:
    Phosphate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Neutral
  • Description:
    di-Sodium hydrogen phosphate solution, 1/15 mol/l
  • M-Clarity quality level:
    MQ100
  • pH value:
    9,2

Specifications

About this item

Buffer stock solution 1/15 mol/l