Flumazenil, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1273808.
- 1273819.
- 1273820.
- 1273841.
- Description:
- Flumazenil
- Flumazenil Related Compound A
- Flumazenil Related Compound B
- Flumazenil Related Compound C
- Synonym(s):
- 8-Fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylic acid
- Ethyl 8-hydroxy-5,6-dihydro-5-methyl-6-oxo-4H-imidazol-[1,5-a][1,4]benzodiazepine-3-carboxylate
- N,N-Dimethylformamide diethyl acetal
- CAS No.:
- 78755-81-4
- 84378-44-9
- 131666-45-0
- 1188-33-6
- Formula:
- C₁₅H₁₄FN₃O₃
- C₇H₁₇NO₂
- UN No.:
- 1993
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1273808.FlumazenilSpecifications:
More
Specifications
Cat. No.1273808.DescriptionFlumazenilSynonym(s)CAS No.78755-81-4FormulaC₁₅H₁₄FN₃O₃UN No. - 1273819.Flumazenil Related Compound AFlumazenil Related Compound AUSP reference standardVial20 mgLog in for availability1 * 20 mg1 520,00 €Specifications:
More
Specifications
Cat. No.1273819.DescriptionFlumazenil Related Compound ASynonym(s)8-Fluoro-5-methyl-6-oxo-5,6-dihydro-4H-benzo[f]imidazo[1,5-a][1,4]diazepine-3-carboxylic acidCAS No.84378-44-9FormulaUN No. - 1273820.Flumazenil Related Compound BFlumazenil Related Compound BUSP reference standardVial20 mgLog in for availability1 * 20 mg1 520,00 €Specifications:
More
Specifications
Cat. No.1273820.DescriptionFlumazenil Related Compound BSynonym(s)Ethyl 8-hydroxy-5,6-dihydro-5-methyl-6-oxo-4H-imidazol-[1,5-a][1,4]benzodiazepine-3-carboxylateCAS No.131666-45-0FormulaUN No. - 1273841.Flumazenil Related Compound CFlumazenil Related Compound CUSP reference standardAmpule500 mgLog in for availability1 * 500 mg1 580,00 €Specifications:
More
Specifications
Cat. No.1273841.DescriptionFlumazenil Related Compound CSynonym(s)N,N-Dimethylformamide diethyl acetalCAS No.1188-33-6FormulaC₇H₁₇NO₂UN No.1993



