Fludeoxyglucose, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1272907.
- 1272918.
- 1272929.
- Description:
- Fludeoxyglucose
- Fludeoxyglucose Related Compound A
- Fludeoxyglucose Related Compound B
- Synonym(s):
- 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
- 2-chloro-2-deoxy-D-glucose
- CAS No.:
- 29702-43-0
- 23978-09-8
- 14685-79-1
- Formula:
- C₆H₁₁FO₅
- C₁₈H₃₆N₂O₆
- C₆H₁₁ClO₅
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1272907.FludeoxyglucoseSpecifications:
More
Specifications
Cat. No.1272907.DescriptionFludeoxyglucoseSynonym(s)CAS No.29702-43-0FormulaC₆H₁₁FO₅ - 1272918.Fludeoxyglucose Related Compound AFludeoxyglucose Related Compound AUSP reference standard15 mgLog in for availability1 * 15 mg1 420,00 €Specifications:
More
Specifications
Cat. No.1272918.DescriptionFludeoxyglucose Related Compound ASynonym(s)4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosaneCAS No.23978-09-8FormulaC₁₈H₃₆N₂O₆ - 1272929.Fludeoxyglucose Related Compound BFludeoxyglucose Related Compound BUSP reference standard10 mgLog in for availability1 * 10 mg1 260,00 €Specifications:
More
Specifications
Cat. No.1272929.DescriptionFludeoxyglucose Related Compound BSynonym(s)2-chloro-2-deoxy-D-glucoseCAS No.14685-79-1FormulaC₆H₁₁ClO₅
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



