Standards for water pollution QC check, residual chlorine, VHG
Catalog # PROOVHG-QWSRCL-2
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:2 mL
- Description:Water analysis standard for residual chlorine
- Type:Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to free and total residual chlorines.
This standard has been prepared as a concentrate and must be diluted prior to analysis. This concentrate is not preserved. Since the diluted standard is not stable, it must be analysed immediately after the concentrate is diluted.
Certifications: Traceable to NIST SRM.