Standards for water pollution QC check, minerals, VHG
Catalog # PROOVHG-QWPMIN-500
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:500 mL
- Description:Water analysis standard for minerals
- Type:Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to minerals.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
Certifications: Traceable to NIST SRM.