ContactUsLinkComponent
Standards for water pollution QC check, mercury, VHG
  PROOVHG-QWPHG-15
 :  
Standards for water pollution QC check, mercury, VHG
  PROOVHG-QWPHG-15
 :  
  • Pk:
    15 mL
  • Description:
    Water analysis standard for mercury, 15 ml
  • Type:
    Reference standard
This solution is intended for use as a quality control check for methods related to mercury.
This standard has been preserved with approximately 1% (v/v) nitric acid and 0,04% (w/v) potassium dichromate. This standard has been prepared as a concentrate and must be diluted prior to analysis.
 : Traceable to NIST SRM.