Order Entry
Belgium
ContactUsLinkComponent
 
Sample changer for KF titration, Stromboli KF
  METN51105200
Sample changer for KF titration, Stromboli KF
  METN51105200

 

  • Description:
    Stromboli KF oven sample changer
  • Model:
    Stromboli KF
  • Power supply:
    100 - 240 VAC
  • Weight:
    5 kg
  • W×D×H:
    250×400×300 mm
  • Pk:
    1

 

 

In combination with a V30 or C30 Karl Fischer titrator the Stromboli sample changer enables up to 14 samples to be titrated in unattended operation. The samples are automatically transferred into a temperature controlled oven from where the evolved water is transferred to the titration cell via an applied gas flow.

  • Controlled by the titrator: Stromboli does not have any operating elements and does not require special installation or configuration
  • Displays the actual and the preset oven temperature in large blue illuminated numerals on the top of the instrument
  • 20 ml sample vials are designed for quick and easy handling

Stromboli is ideal for accurate water determination of solid or viscous samples like:
- Substances that release water only at elevated temperatures such as plastics in powder or granular form
- Substances that cause side reactions with Karl Fischer reagent such as oxidizing inorganic salts
- Substances that cause problems in the titration vessel due to their consistency such as fibrous or pasty substances
- Substances that are poorly soluble or insoluble such as wool, dough, tar or coal

Application recommendations:
- The water content should be at least 500 µg per sample
- As purge gas air or nitrogen can be used. For samples that decompose with air at high temperatures, the use of nitrogen is recommended

Temperature range: 50 - 300 °C
Temperature accuracy: ±3 °C