Saisie de commande
Belgium
ContactUsLinkComponent
Standards for water pollution QC check, sample demand, VHG
Standards for water pollution QC check, sample demand, VHG
  PROOVHG-QWPDEM-15
 :  
Standards for water pollution QC check, sample demand, VHG
  PROOVHG-QWPDEM-15
 :  
  • Cdt:
    15 mL
  • Désignation:
    Water analysis standard for BOD, CBOD, COD, TOC
  • Type:
    Reference standard
This solution is intended for use as a quality control check for methods related to sample demands including BOD, CBOD, COD, TOC.
This standard has been prepared as a concentrate and must be diluted prior to analysis. This concentrate is preserved with approximately 1% (v/v) hydrochloric acid.
 : Traceable to NIST SRM.