ContactUsLinkComponent
Standards for water pollution QC check, complex nutrients, VHG
Standards for water pollution QC check, complex nutrients, VHG
Catalog # PROOVHG-QWPCNUT-15
CAS Number:  
Standards for water pollution QC check, complex nutrients, VHG
Catalog # PROOVHG-QWPCNUT-15
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    Water analysis standard for complex nutrients
  • Type:
    Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to complex nutrients.
This standard has been prepared as a concentrate and must be diluted prior to analysis. This concentrate is preserved with approximately 1% (v/v) hydrochloric acid.
Certifications: Traceable to NIST SRM.