Orders LinkContactUsLinkComponent
Tissue grinders, CS1, KIMBLE®
Tissue grinders, CS1, KIMBLE®
Supplier:  DWK Life Sciences
Certificates
Tissue grinders, CS1, KIMBLE®
Tissue grinders, CS1, KIMBLE®
2 Options Available Below
Product Details & Documents
CS1 tissue grinders are suitable for standard grinding, tissues are ground within a sealed container to minimise the risk of personal contact.
  • Narrow pestles eliminate the potential of sample overflow
  • Convenient and easy-to-use
  • Gamma sterilised in an easy-open Tyvek® pack
The molded-in abrasive surface on the pestle tip leaves no sediment to obstruct sample examination. Tubes are made of PP, pestles are 30% glass-filled PP and caps are HDPE.
 : Each tissue grinder is supplied with a fully assembled pestle, a conical sample tube with solid top cap, an adhesive identification label and an instruction card.
Product Family Options
Product Information