Sodium-2-propylvalerate ≥98% (by HPLC), Millipore®
Catalog # 676380-5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:166.20 g/mol
:Packaged under inert gas.
Storage Temperature:Refrigerator
CAS Number:1069-66-5
Formula:C₈H₁₅O₂Na
MDL Number:MFCD00078604