Order Entry
Belgium
ContactUsLinkComponent
Storage boxes, SP Bel-Art
Storage boxes, SP Bel-Art
Catalog # BELAF166140000
undefined
Storage boxes, SP Bel-Art
Catalog # BELAF166140000

Specifications

  • Colour:
    Crystal clear
  • Description:
    24 compartments
  • Seq:
    2
  • W×D×H:
    333×229×58 mm
  • Pk:
    1

Specifications

About this item

This styrene plastic compartment box is designed to organise and store 35 mm slides, hardware, corks, stoppers and other lab items.

  • Crystal-clear, they afford full visibility of contents
  • Tight-fitting, hinged lid keeps objects contained
  • Not recommended below 0 °C