Orders LinkContactUsLinkComponent
Push button safety covers
Push button safety covers
Supplier:   Brady
Push button safety covers
Supplier:  Brady
Certificates
Push button safety covers
Push button safety covers
2 Options Available Below
Product Details & Documents
These devices made from metal and clear PC guard against unauthorised operation or accidental activation.
  • Clear Lexan cover with hinged door can be kept open for easy access when not in use
  • For use with button style IEC 22,5 mm or NEMA 30,5 mm
Product Family Options
Product Information
  • Type
    W×D×H
    Availability
    Price
  • Push button safety cover, for use with button style IEC 22,5 mm
    37×34,1×58,4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • Push button safety cover, for use with button style NEMA 30,5 mm, incl. white legend plate
    39,87×49,02 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 110,00 €
    110,00 €
    1 * 1 ST
       110,00 €