Elemental analysis standards, total kjeldahl nitrogen (TKN) standards, VHG
Catalog # PROOVHG-TOTKJN1K-1
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Concentration:1000 mg/l in 1% HCl
- Pk:100 mL
- Description:Total Kjeldahl nitrogen standard
Specifications
About this item
This standard solution is intended for use as a calibration standard for total Kjeldahl nitrogen (TKN). It is prepared gravimetrically to contain nominally 1000 mg of total Kjeldahl nitrogen per liter of solution using analytical grade raw materials and 18 megohm deionised water. The certified concentration is the mean of replicate measurements. The uncertainty is calculated as ±2s/√n.
Certifications: Traceable to NIST and SRM.