Selenium standards, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
This standard solution is intended for use as a calibration and reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.
Certifications: Accredited to ISO 9001 and ISO/IEC 17025. Traceable to NIST and SRM.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together