Order Entry
Belgium
ContactUsLinkComponent
Multi-element standards, comprehensive mix A, VHG
Multi-element standards, comprehensive mix A, VHG
Catalog # PROOVHG-SM80C-500
CAS Number:  
Multi-element standards, comprehensive mix A, VHG
Catalog # PROOVHG-SM80C-500
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    500 mL
  • Description:
    Comprehensive Mix A standard

Specifications

About this item

This standard solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).

Multi element standards for Ag, Al, As, Ba, Bi, Ca, Cd, Ce, Dy, Er, Eu, Ga, Gd, Ho, La, Lu, Mg, Na, Nd, P, Pb, Pr, Rb, Sc, Se, Sm, Sr, Tb, Th, Tl, Tm, U, Y, Yb at 10 µg/ml in 40% aqua regia.

Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.