Orders LinkContactUsLinkComponent
Multi-element standards, common and transition elements, VHG
Multi-element standards, common and transition elements, VHG
Certificates
Multi-element standards, common and transition elements, VHG
2 Options Available Below
Product Details & Documents
This standard solution is intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi elements standards for Ag, Al, As, Ba, Be, Bi, Cd, Cr, Co, Cu, Fe, Li, Mn, Mo, Ni, Pb, Sb, Se, Sr, Tl, V, Zn at 100 µg/ml and Ca, K, Mg, Na at 1000 µg/ml in 0,2% HF.
 : Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Product Family Options
Product Information
  • Pk Info
    Pk
    Availability
    Price
  • PROOVHG-SM75B-100
    Common and transition element standards
    Common and transition element standards
    100 ml
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 698,00 €
    698,00 €
    1 * 100 mL
       698,00 €
  • PROOVHG-SM75B-500
    Common and transition element standards
    Common and transition element standards
    500 ml
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 1 400,00 €
    1 400,00 €
    1 * 500 mL
       1 400,00 €