Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-element standards, major cation mix, VHG

Multi-element standards, major cation mix, VHG

Major cation mix, 1000 mg/L, 5% HNO₃, 500 ml, 500 mL
Catalog # PROOVHG-SM16-500
Multi-element standards, major cation mix, VHG
Catalog # PROOVHG-SM16-500
CAS Number:  
Multi-element standards, major cation mix, VHG

Major cation mix, 1000 mg/L, 5% HNO₃, 500 ml, 500 mL

CAS Number:  
Price
712,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).

Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.

Specifications

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HNO₃
  • Pk:
    500 mL
  • Description:
    Major cation mix
  • Pk Info:
    500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.