Empty racks for tipfill system, Qualitix®
Supplier: Socorex Isba
Jump to:
About this item
These empty racks are designed to be used with Qualitix® tipfill packs.
- Racks have smooth rounded edges and ergonomic snap latch that won’t catch gloves
- Transparent lid enables instant view of content
- Empty racks are re-usable and autoclavable
2 Options Available Below
Product Details & Documents
These empty racks are designed to be used with Qualitix® tipfill packs.
- Racks have smooth rounded edges and ergonomic snap latch that won’t catch gloves
- Transparent lid enables instant view of content
- Empty racks are re-usable and autoclavable
Product Family Options
Product Information
- AvailabilityPrice
- SOCO3070.0210EREmpty rack for 10/200 µl tips, to be used with tipfill systemEmpty rack for 10/200 µl tips, to be used with tipfill systemLog in for availability1 * 10 ST37,70 €
- SOCO3070.1000EREmpty rack for 1000 µl tips, to be used with tipfill system