Order Entry
Belgium
ContactUsLinkComponent
Test tube racks, In-Line, SP Bel-Art
Test tube racks, In-Line, SP Bel-Art
Catalog # BELAF185270000
undefined
Test tube racks, In-Line, SP Bel-Art
Catalog # BELAF185270000

Specifications

  • For tube Ø:
    20 - 25 mm
  • No. of Holes:
    6 (6×1)
  • W×D×H:
    241×76×92 mm
  • Pk:
    1

Specifications

About this item

For high-visibility, six-place high-density polyethylene rack is end-supported by two columns.

  • Top plate has 25 mm diameter holes
  • Base has corresponding hemispherical wells
  • Includes six vertical pins for tube drying
  • Easy snap together assembly