Elemental analysis standards, rare earth and geo elements standard, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi element standards for Ba, Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Rb, Sc, Sm, Sr, Tb, Th, Tm, U, Y, Yb at 100 µg/ml in 5% HNO₃.
Certifications: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together