Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
Conductivity Sensors
Accessories for conductivity sensors
Accessories for conductivity sensors
Accessories for conductivity sensors
Catalog # KNICZU0298
Supplier:
KNICK
Expand image
Accessories for conductivity sensors
Catalog # KNICZU0298
Supplier:
KNICK
Specifications
Description:
Adapter for connecting the SE 202 and SE 204 sensors to the 703 laboratory conductivity meter
Pk:
1
Specifications
About this item
Product Details
Product Details