Yttrium standard solution, for ICP, in HNO₃, CRM, VHG
These standard solution are intended for use as a certified reference material and calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF) and other techniques for elemental analysis.
: Traceable to NIST, SRM.
- Environmentally Preferable:
Frequently Bought Together