Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Corrosive
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    49.5 - 50.5 %
  • Ammonium Hydroxide Precipitate:
    max. 0.01 %
  • Chloride (Cl):
    max. 0.002 %
  • Potassium (K):
    max. 0.01 %
  • Heavy Metals (as Ag):
    max. 0.001 %
  • Sodium Carbonate (Na2CO3):
    max. 0.3 %
  • Sulfate (SO4):
    max. 0.001 %
  • Iron (Fe):
    max. 5
  • Nickel (Ni):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5
  • Phosphate (PO4):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together