Order Entry
Belgium
ContactUsLinkComponent
 
Spill containment tray with grid, SP Bel-Art
  BELAF246760003
undefined
Spill containment tray with grid, SP Bel-Art
  BELAF246760003

 

  • Width:
    308 mm
  • Description:
    Spill containment tray with grid, 7 L
  • Height:
    121 mm
  • Length:
    365 mm
  • Pk:
    1

 

 

This large, deep, polypropylene plastic tray protects users by capturing accidental hazardous spills.

  • High chemical resistance
  • Epoxy coated wire support grid holds numerous vessels or racks of test tubes
  • Steam autoclavable at 121 °C