Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Petroleum Industry Standards
Biodiesel standard, sulphur in mineral oil, CONOSTAN
Biodiesel standard, sulphur in mineral oil, CONOSTAN
Biodiesel standard, sulphur in mineral oil, CONOSTAN
Catalog # PROOCON150-400-010
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Biodiesel standard, sulphur in mineral oil, CONOSTAN
Catalog # PROOCON150-400-010
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Concentration:
25 µg/g
Pk:
100 g
Description:
Sulphur in mineral oil
Specifications
About this item
Product Details
Product Details