Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
Supplier:   Sampling Systems
Measuring scoops
Supplier:  Sampling Systems
Certificates
Measuring scoops
Measuring scoops
5 Options Available Below
Product Details & Documents
These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.
  • Crevice-free
  • Polished
Product Family Options
Product Information
  • Capacity
    Length
    Ø
    Availability
    Price
  • 1 ml
    186 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 91,10 €
    91,10 €
    1 * 1 ST
       91,10 €
  • 5 ml
    193 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
  • 10 ml
    193 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 138,00 €
    138,00 €
    1 * 1 ST
       138,00 €
  • 25 ml
    199 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 148,00 €
    148,00 €
    1 * 1 ST
       148,00 €
  • 50 ml
    199 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 176,00 €
    176,00 €
    1 * 1 ST
       176,00 €