Order Entry
Belgium
ContactUsLinkComponent
n-Heptane ≥99.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Flammable
Health
Irritant
Environment
n-Heptane ≥99.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9177.1000
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Heptane ≥99.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9177.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99.0 %
  • Residue after Evaporation:
    max. 2 ppm
  • Substances Darkened by H2SO4:
    passes test
  • Water (by KF, coulometric):
    max. 0.01 %
  • Measured at Emission Maximum for Solvent Impurities:
    max. 1.0
  • Measured at 450 nm:
    max. 0.2
  • at 210 nm:
    max. 0.40
  • at 220 nm:
    max. 0.10
  • at 254-400 nm:
    max. 0.01
  • UV Cut-off, nm:
    max. 197

Specification Test Results

About this item

 :
 :100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544