Order Entry
Belgium
ContactUsLinkComponent
Trifluoroacetic acid ≥99.5%, BAKER ANALYZED® HPLC, J.T.Baker®
Trifluoroacetic acid ≥99.5%, BAKER ANALYZED® HPLC, J.T.Baker®
Catalog # 9470.0010
Supplier:  Avantor
CAS Number:  
undefined
Trifluoroacetic acid ≥99.5%, BAKER ANALYZED® HPLC, J.T.Baker®
Catalog # 9470.0010
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass vial
  • Pk:
    10 Vial
  • Seq:
    2076
  • Pk Info:
    10x 1 ml

Specifications

Specification Test Results

  • Assay:
    min. 99.5 %
  • Heavy Metals (as Pb):
    max. 10 ppm
  • Residue after Evaporation:
    max. 10 ppm
  • at 215 nm:
    max. 0.05
  • Iron (Fe):
    max. 5
  • at 230 nm:
    max. 0.15
  • at 254 nm:
    max. 0.01
  • UV Cut-off, nm:
    max. 210

Specification Test Results

About this item

 :
 :114.02 g/mol
UN:2699
ADR:8,I
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169