Order Entry
Belgium
ContactUsLinkComponent
n-Hexane, anhydrous (max. 0.003% H₂O)
Flammable
Health
Toxic
Environment
n-Hexane, anhydrous (max. 0.003% H₂O)
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Toxic
Environment
n-Hexane, anhydrous (max. 0.003% H₂O)
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay (GC, on anhydrous basis):
    99-100 %
  • Color (APHA, Pt-Co):
    0-10
  • Water (KF):
    0-0.003 % w/w
  • Acidity (as Acetic acid):
    0-0.001 %
  • Residue after evaporation:
    0-0.0005 % w/w

Specification Test Results

Related Information

Frequently Bought Together