ContactUsLinkComponent
tert-Butyl methyl ether, anhydrous (max. 0.005% H₂O)
Flammable
Irritant
tert-Butyl methyl ether, anhydrous (max. 0.005% H₂O)
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
tert-Butyl methyl ether, anhydrous (max. 0.005% H₂O)
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale
Specification Test Results
  • Assay (GC, on anhydrous basis):
    99-100 %
  • Acidity:
    0-0.001 meq/g
  • Peroxides (as H202):
    0-0.0005 %
  • Residue after evaporation:
    0-0.001 % w/w
  • Water (KF) :
    0-0.005 % w/w