Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Triamcinolone, Ph.Eur., RS
Organic reference standard, Triamcinolone, Ph.Eur., RS
Catalog # EDQMT1899990
Supplier:  EDQM
CAS Number:  
Organic reference standard, Triamcinolone, Ph.Eur., RS
Catalog # EDQMT1899990
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    150 mg
  • Standards:
    Reference standard
  • CAS No.:
    124-94-7
  • Description:
    Triamcinolone
  • Formula:
    C₂₁H₂₇FO₆
  • Storage temperature:
    Ambient
  • Description:
    Triamcinolone

Specifications

About this item

  • In Monograph 1376
  • Solubility in/miscible with DMSO and methanol