Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Tianeptine for system suitability, Ph.Eur., RS
Organic reference standard, Tianeptine for system suitability, Ph.Eur., RS
Catalog # EDQMY0000152
Supplier:  EDQM
CAS Number:  
Organic reference standard, Tianeptine for system suitability, Ph.Eur., RS
Catalog # EDQMY0000152
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    30 mg
  • Standards:
    Reference standard
  • CAS No.:
    30123-17-2
  • Description:
    Tianeptine for system suitability
  • Formula:
    C₂₁H₂₄ClN₂NaO₄S
  • Storage temperature:
    Ambient
  • Description:
    Tianeptine for system suitability

Specifications

About this item

Filled under inert atmosphere protected from humidity.

  • In Monograph 2022
  • Solubility in water: 100 - 1000 g/l