Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Tiamulin hydrogen fumarate, Ph.Eur., RS
Organic reference standard, Tiamulin hydrogen fumarate, Ph.Eur., RS
Catalog # EDQMY0000333
Supplier:  EDQM
CAS Number:  
Organic reference standard, Tiamulin hydrogen fumarate, Ph.Eur., RS
Catalog # EDQMY0000333
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    500 mg
  • Standards:
    Reference standard
  • CAS No.:
    55297-96-6
  • Description:
    Tiamulin hydrogen fumarate
  • Formula:
    C₃₂H₅₁NO₈S
  • Storage temperature:
    Ambient
  • Description:
    Tiamulin hydrogen fumarate

Specifications

About this item

  • Pharmaceutical applications