Organic reference standard, Simvastatin, Ph.Eur., RS
Catalog # EDQMS0650000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:200 mg
- Standards:Reference standard
- CAS No.:79902-63-9
- Description:Simvastatin
- Formula:C₂₅H₃₈O₅
- Storage temperature:Ambient
- Description:Simvastatin
Specifications
About this item
- In Monograph 1563
- Soluble in/miscibility with ethyl acetate and methanol
- Pharmaceutical applications