Organic reference standard, Pidolic acid, Ph.Eur., RS
Catalog # EDQMY0000123
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:125 mg
- Standards:Reference standard
- CAS No.:98-79-3
- Description:Pidolic acid
- Formula:C₅H₇NO₃
- Storage temperature:Ambient
- Description:Pidolic acid
Specifications
About this item
- In Monograph 1619
- Pharmaceutical applications