Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, L-Arabinitol, Ph.Eur., RS
Organic reference standard, L-Arabinitol, Ph.Eur., RS
Catalog # EDQMA1269000
Supplier:  EDQM
CAS Number:  
Organic reference standard, L-Arabinitol, Ph.Eur., RS
Catalog # EDQMA1269000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    7643-75-6
  • Description:
    L-Arabinitol
  • Formula:
    C₅H₁₂O₅
  • Description:
    L-Arabinitol

Specifications

About this item

  • Method: Monograph only
  • Excipient product type
  • Pharmaceutical industry applications