Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Eicosapentaenoic acid ethyl ester, Ph.Eur., RS
Organic reference standard, Eicosapentaenoic acid ethyl ester, Ph.Eur., RS
Catalog # EDQME0085000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Eicosapentaenoic acid ethyl ester, Ph.Eur., RS
Catalog # EDQME0085000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    Reference standard
  • CAS No.:
    73310-10-8
  • Description:
    Eicosapentaenoic acid ethyl ester
  • Formula:
    C₂₂H₃₄O₂
  • Storage temperature:
    Ambient
  • Description:
    Eicosapentaenoic acid ethyl ester

Specifications

About this item

  • In Monograph 20429
  • Pharmaceutical applications