Order Entry
Belgium
ContactUsLinkComponent
 
Organic reference standard, Cyclobenzaprine hydrochloride, Ph.Eur., RS
Catalog # EDQMC3200000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Cyclobenzaprine hydrochloride, Ph.Eur., RS
Catalog # EDQMC3200000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    6202-23-9
  • Description:
    Cyclobenzaprine hydrochloride
  • Formula:
    C₂₀H₂₂ClN
  • Storage temperature:
    Ambient
  • Description:
    Cyclobenzaprine hydrochloride

Specifications

About this item

  • In Monographs 0408 and 0464