Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Cortisone acetate, Ph.Eur., RS
Organic reference standard, Cortisone acetate, Ph.Eur., RS
Catalog # EDQMC2800000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Cortisone acetate, Ph.Eur., RS
Catalog # EDQMC2800000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    250 mg
  • Standards:
    Reference standard
  • CAS No.:
    50-04-4
  • Description:
    Cortisone acetate
  • Formula:
    C₂₃H₃₀O₆
  • Storage temperature:
    Ambient
  • Description:
    Cortisone acetate

Specifications

About this item

  • In Monographs 0321, 0736, and 0767
  • Solubility in water: less than 1 g/l