Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, colchicine, Ph.Eur., RS
Organic reference standard, colchicine, Ph.Eur., RS
Catalog # EDQMC2580000
Supplier:  EDQM
CAS Number:  
Organic reference standard, colchicine, Ph.Eur., RS
Catalog # EDQMC2580000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    15 mg
  • Standards:
    Reference standard
  • CAS No.:
    64-86-8
  • Description:
    Colchicine
  • Formula:
    C₂₂H₂₅NO₆
  • Storage temperature:
    Ambient
  • Description:
    Colchicine

Specifications

About this item

  • In Monograph 0758