Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, ciprofloxacin, Ph.Eur., RS
Organic reference standard, ciprofloxacin, Ph.Eur., RS
Catalog # EDQMY0000198
Supplier:  EDQM
CAS Number:  
Organic reference standard, ciprofloxacin, Ph.Eur., RS
Catalog # EDQMY0000198
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    Reference standard
  • CAS No.:
    85721-33-1
  • Description:
    Ciprofloxacin
  • Formula:
    C₁₇H₁₈FN₃O₃
  • Storage temperature:
    Ambient
  • Description:
    Ciprofloxacin

Specifications

About this item

  • In Monograph 1089
  • Soluble in/miscibility with hot DMSO and hot methanol