Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Chlorhexidine, Ph.Eur., RS
Organic reference standard, Chlorhexidine, Ph.Eur., RS
Catalog # EDQMC1510000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Chlorhexidine, Ph.Eur., RS
Catalog # EDQMC1510000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    55-56-1
  • Description:
    Chlorhexidine
  • Formula:
    C₂₂H₃₀Cl₂N₁₀
  • Storage temperature:
    Ambient
  • Description:
    Chlorhexidine

Specifications

About this item

  • In Monograph 0658