Organic reference standard, Cefadroxil, Ph.Eur., RS
EDQMC0650000
: EDQM
:
- Pack type:Vial
- Pk:250 mg
- Standards:Reference standard
- CAS No.:50370-12-2
- Description:Cefadroxil
- Formula:C₁₆H₁₇N₃O₅S
- Storage temperature:Ambient
- Description:Cefadroxil
- Solubility in water and methanol
- Pharmaceutical industry applications