Organic reference standard, Betamethasone, Ph.Eur., RS
Catalog # EDQMB1000000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:378-44-9
- Description:Betamethasone
- Formula:C₂₂H₂₉FO₅
- Storage temperature:Ambient
- Description:Betamethasone
Specifications
About this item
- In Monographs 0312,0354,0388, and 0811
- Solubility in water: less than 0,063 g/l