Order Entry
Belgium
ContactUsLinkComponent
Organic reference standard, Aspartame impurity A 2-(5-Benzyl-3,6-dioxopiperazin-2-yl) acetic acid , Ph.Eur., RS
Organic reference standard, Aspartame impurity A 2-(5-Benzyl-3,6-dioxopiperazin-2-yl) acetic acid , Ph.Eur., RS
Catalog # EDQMA1321000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Aspartame impurity A 2-(5-Benzyl-3,6-dioxopiperazin-2-yl) acetic acid , Ph.Eur., RS
Catalog # EDQMA1321000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    Reference standard
  • CAS No.:
    5262-10-2
  • Description:
    Aspartame impurity A 2-(5-Benzyl-3,6-dioxopiperazin-2-yl) acetic acid
  • Formula:
    C₁₃H₁₄N₂O₄
  • Storage temperature:
    +5°C ± 3°C
  • Description:
    Aspartame impurity A 2-(5-Benzyl-3,6-dioxopiperazin-2-yl) acetic acid

Specifications

About this item

  • Method: Monograph only
  • Product type: Impurity
  • Pharmaceutical industry applications