Organic reference standard, 4-Aminobenzoic acid, Ph.Eur., RS
Catalog # EDQMY0000173
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:60 mg
- Standards:Reference standard
- CAS No.:150-13-0
- Description:4-Aminobenzoic acid
- Formula:C₇H₇NO₂
- Storage temperature:Ambient
- Description:4-Aminobenzoic acid
Specifications
About this item
- In Monograph 1687
- Solubility in water: 8 g/l at 25°C