ContactUsLinkComponent
Pipette stand, Poxygrid®, SP Bel-Art
Pipette stand, Poxygrid®, SP Bel-Art
Catalog # BELAH189620000
undefined
Pipette stand, Poxygrid®, SP Bel-Art
Catalog # BELAH189620000
Specifications
  • For:
    10 standard pipettes (Finnpette®, Socorex®, Eppendorf® pipettes); 2 pipettes with profile like Drummond Pipette-Aid®
  • Colour:
    Blue
  • Seq:
    01
  • W×D×H:
    229×127×171 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This epoxy-coated aluminum rack stores pipettes and protects pipette tips.
  • Holds up to 10 round shape pipettors such as Finnpipette®, Socorex®, Eppendorf® and other popular brands; two end holes fit Pipette-Aid® pipette fillers
  • Two stepped rows of 5 each
  • Second lower support row has smaller holes to accept tapered ends of the pipettors to keep them vertical
  • Quick and easy access for more efficient work; no more searching, fumbling or breakage