Orders LinkContactUsLinkComponent
VWR®, Digital Timer, One Channel
VWR®, Digital Timer, One Channel
White, 0,01 %
Catalog # 609-0224
Supplier:   VWR Collection
VWR®, Digital Timer, One Channel
Catalog # 609-0224
Supplier:  VWR Collection
VWR®, Digital Timer, One Channel
VWR®, Digital Timer, One Channel
White, 0,01 %
Price
70,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
This single-channel timer can be programmed in countdown or count up mode.
  • Timing capacity: 99 minutes, 59 seconds
  • Resolution is 1 second
  • Alarm and memory function
  • Big digit display
Delivery information: Supplied with magnet, clip and stand.
Specifications
  • Colour:
    White
  • Accuracy:
    0,01 %